trans-4-Aminocyclohexanol 98%

Supplier: Ambeed
Danger

A775896-1000G A775896-100G A775896-10G A775896-25G A775896-500G A775896-5G
76791-532EA 129 USD
76791-532 76791-534 76791-536 76791-538 76791-540 76791-542
trans-4-Aminocyclohexanol 98%
trans-4-Aminocyclohexanol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MDL Number: MFCD00067698
CAS Number: 27489-62-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR