N-Boc-4-iodo-2-methylaniline 97+%

Supplier: Ambeed
Warning

A775623-250MG A775623-5G
76791-528EA 25.47 USD
76791-528 76791-530
N-Boc-4-iodo-2-methylaniline 97+%
N-Boc-4-iodo-2-methylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₆INO₂
MW: 333.16 g/mol
MDL Number: MFCD08703145
CAS Number: 666746-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR