Tris(3-aminopropyl)amine 98%

Supplier: Ambeed
Danger

Synonyms: N1,N1-Bis(3-aminopropyl)propane-1,3-diamine, 3,3',3''-Nitrilotris(propylamine)

A775187-100MG A775187-250MG
76791-524EA 29.71 USD
76791-524 76791-526
Tris(3-aminopropyl)amine 98%
Tris(3-aminopropyl)amine

Precaución: for Research Use Only

Formula: C₉H₂₄N₄
MW: 188.32 g/mol
MDL Number: MFCD00191593
CAS Number: 4963-47-7
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR