6-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(Pyrrolidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(Pyrrolidino)pyridine-5-boronic acid pinacol ester
A775085-10G
A775085-1G
A775085-250MG
A775085-5G
76791-518EA
165
USD
76791-518
76791-520
76791-522
77670-076
6-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester 97%
6-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₂₃BN₂O₂ |
MDL Number:
MFCD06798273 CAS Number: 933986-97-1 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...