3-Bromophenanthrene 98% (by HPLC)

Supplier: Ambeed
Danger

A773252-1G A773252-250MG A773252-5G
76791-498EA 47 USD
76791-498 76791-500 76791-502
3-Bromophenanthrene 98% (by HPLC)
3-Bromophenanthrene

Precaución: for Research Use Only

Formula: C₁₄H₉Br
MDL Number: MFCD06797074
CAS Number: 715-50-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR