Methyl-2-(p-tolyl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(4-Methylphenyl)benzoate, 4'-Methylbiphenyl-2-carboxylic Acid Methyl Ester, Methyl 4'-methyl-[1,1'-biphenyl]-2-carboxylate, Methyl 4'-Methylbiphenyl-2-carboxylate, 2-(p-Tolyl)benzoic Acid Methyl Ester

A773072-100G A773072-1G A773072-25G A773072-500G A773072-5G
76791-488EA 448.77 USD
76791-488 76791-490 76791-492 76791-494 76791-496
Methyl-2-(p-tolyl)benzoate 98%
Methyl-2-(p-tolyl)benzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
MW: 226.28 g/mol
MDL Number: MFCD03453661
CAS Number: 114772-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR