2-Bromo-3,5-dichloroaniline 95%

Supplier: Ambeed
Warning

A770830-1G A770830-250MG A770830-5G
76791-468EA 60.48 USD
76791-468 76791-470 76791-472
2-Bromo-3,5-dichloroaniline 95%
2-Bromo-3,5-dichloroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrCl₂N
MW: 240.9100 g/mol
MDL Number: MFCD09999825
CAS Number: 1211214-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR