3-Chloro-2-nitroanisole 96%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-3-methoxy-2-nitrobenzene

A770393-1G A770393-250MG
76791-464EA 73.21 USD
76791-464 76791-466
3-Chloro-2-nitroanisole 96%
3-Chloro-2-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
MDL Number: MFCD14455916
CAS Number: 5472-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR