4-[2-(trans-4-Pentylcyclohexyl)ethyl]cyclohexanone 95+%

Supplier: Ambeed
Warning

Synonyms: trans-4-[2-(4-Pentylcyclohexyl)ethyl]cyclohexanone, 4-[2-(trans-4-Amylcyclohexyl)ethyl]cyclohexanone

A768128-1G A768128-5G
76791-460EA 23 USD
76791-460 76791-462
4-[2-(trans-4-Pentylcyclohexyl)ethyl]cyclohexanone 95+%
4-[2-(trans-4-Pentylcyclohexyl)ethyl]cyclohexanone

Precaución: for Research Use Only

Formula: C₁₉H₃₄O
MW: 278.48 g/mol
MDL Number: MFCD14636435
CAS Number: 121040-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR