tert-Butyldimethylsilyl-(S)-glycidyl ether 95%

Supplier: Ambeed
Warning

A766635-1G A766635-5G A766635-100G
76791-450EA 12 USD
76791-450 76791-452 77873-218
tert-Butyldimethylsilyl-(S)-glycidyl ether 95%
tert-Butyldimethylsilyl-(S)-glycidyl ether

Precaución: for Research Use Only

Formula: C₉H₂₀O₂Si
MDL Number: MFCD01862148
CAS Number: 123237-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR