trans-2-Pentenoic acid 97%

Supplier: Ambeed
Danger

Synonyms: (E)-Pent-2-enoic acid, 2-Pentenoic acid

A765691-25G A765691-5G
76791-440EA 75.33 USD
76791-440 76791-442
trans-2-Pentenoic acid 97%
trans-2-Pentenoic acid

Precaución: for Research Use Only

Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00002704
CAS Number: 13991-37-2
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR