6-Chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A765084-1G A765084-250MG
76791-436EA 216.43 USD
76791-436 77670-058
6-Chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid 98%
6-Chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
MDL Number: MFCD09878690
CAS Number: 800402-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR