6-Morpholinopyrimidin-4-amine 98%

Supplier: Ambeed
Warning

A764495-250MG A764495-5G
76791-432EA 17 USD
76791-432 76791-434
6-Morpholinopyrimidin-4-amine 98%
6-Morpholinopyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₈H₁₂N₄O
MDL Number: MFCD09999228
CAS Number: 96225-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR