Tetraethylene glycol dimethacrylate 95% stabilized

Supplier: Ambeed
Warning

Synonyms: ((Oxybis(ethane-2,1-diyl))bis(oxy))bis(ethane-2,1-diyl) bis(2-methylacrylate)

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A762935-100G A762935-25G A762935-5G
76791-420EA 74.27 USD
76791-420 76791-422 76791-424
Tetraethylene glycol dimethacrylate 95% stabilized
Tetraethylene glycol dimethacrylate

Precaución: for Research Use Only

Formula: C₁₆H₂₆O₇
MW: 330.38 g/mol
MDL Number: MFCD00014932
CAS Number: 109-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR