(2R,4R)-tert-Butyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A759392-100MG A759392-1G A759392-250MG
76791-404EA 13.8 USD
76791-404 76791-406 76791-408
(2R,4R)-tert-Butyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate 98%
(2R,4R)-tert-Butyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.26 g/mol
MDL Number: MFCD11617834
CAS Number: 141850-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR