5-Chloro-2-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A758920-1G A758920-250MG A758920-25G A758920-5G
76791-400EA 47 USD
76791-400 76791-402 77670-046 77670-048
5-Chloro-2-methylbenzaldehyde 97%
5-Chloro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO
MDL Number: MFCD11111042
CAS Number: 58966-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR