Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')iron(III) 98%

Supplier: Ambeed
Warning

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), Fe(TMHD)3

A755474-100MG A755474-1G A755474-250MG A755474-5G A755474-25G
76791-380EA 8 USD
76791-380 76791-382 76791-384 76791-386 77873-195
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')iron(III) 98%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')iron(III)

Precaución: for Research Use Only

Formula: C₃₃H₅₇FeO₆
MDL Number: MFCD00067465
CAS Number: 14876-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR