6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A754044-100MG A754044-250MG
76791-354EA 44.56 USD
76791-354 76791-356
6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridin-2(1H)-one 95%
6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD11518944
CAS Number: 601514-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR