2-Bromo-6-cyanopyrazine 98%

Supplier: Ambeed
Warning

A753774-1G A753774-250MG A753774-5G
76791-348EA 67.9 USD
76791-348 76791-350 76791-352
2-Bromo-6-cyanopyrazine 98%
2-Bromo-6-cyanopyrazine

Precaución: for Research Use Only

Formula: C₅H₂BrN₃
MW: 183.9900 g/mol
MDL Number: MFCD11109800
CAS Number: 859064-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR