(R)-(-)-2-Hydroxy-2-phenylethyl-p-toluenesulfonate 98%

Supplier: Ambeed
Warning

A752502-1G A752502-5G
76791-344EA 20 USD
76791-344 76791-346
(R)-(-)-2-Hydroxy-2-phenylethyl-p-toluenesulfonate 98%
(R)-(-)-2-Hydroxy-2-phenylethyl-p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₄S
MDL Number: MFCD00236055
CAS Number: 40434-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR