Methyl-5-bromo-1H-indole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromo-1H-indole-2-carboxylate

A750286-1G A750286-250MG A750286-5G
76791-326EA 45.62 USD
76791-326 76791-328 76791-330
Methyl-5-bromo-1H-indole-2-carboxylate 98%
Methyl-5-bromo-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MW: 254.09 g/mol
MDL Number: MFCD01854034
CAS Number: 210345-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR