7-(2-Hydroxy-3-((2-hydroxyethyl)(methyl)amino)propyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione nicotinate 98%

Supplier: Ambeed
Warning

A748220-100MG A748220-10MG A748220-1G A748220-1MG A748220-50MG
76791-314EA 65 USD
76791-314 76791-316 76791-318 76791-320 76791-322
7-(2-Hydroxy-3-((2-hydroxyethyl)(methyl)amino)propyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione nicotinate 98%
7-(2-Hydroxy-3-((2-hydroxyethyl)(methyl)amino)propyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione nicotinate

Precaución: for Research Use Only

Formula: C₁₉H₂₆N₆O₆
MW: 434.46 g/mol
MDL Number: MFCD00058342
CAS Number: 437-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR