1-Carbobenzoxy-3-piperidinecarboxylic acid 97%

Supplier: Ambeed
Warning

A748145-10G A748145-1G A748145-5G A748145-25G A748145-100G A748145-500G
76791-308EA 10 USD
76791-308 76791-310 76791-312 77670-024 77873-190 77873-191
1-Carbobenzoxy-3-piperidinecarboxylic acid 97%
1-Carbobenzoxy-3-piperidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₄
MDL Number: MFCD02931282
CAS Number: 78190-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR