Vinyltrimethylsilane 97%

Supplier: Ambeed
Danger

Synonyms: Trimethyl(vinyl)silane

A746932-10G A746932-1G A746932-25G
76791-298EA 21.22 USD
76791-298 76791-300 76791-302
Vinyltrimethylsilane 97%
Vinyltrimethylsilane

Precaución: for Research Use Only

Formula: C₅H₁₂Si
MW: 100.24 g/mol
MDL Number: MFCD00008606
CAS Number: 754-05-2
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR