trans-1,2-Bis(diphenylphosphino)ethylene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(E)-1,2-Bis(diphenylphosphino)ethene
A746473-1G
A746473-250MG
A746473-25G
A746473-5G
76791-290EA
95.49
USD
76791-290
76791-292
76791-294
76791-296
trans-1,2-Bis(diphenylphosphino)ethylene 98%
trans-1,2-Bis(diphenylphosphino)ethylene
Precaución: for Research Use Only
Formula:
C₂₆H₂₂P₂ MW: 396.41 g/mol |
MDL Number:
MFCD00063219 CAS Number: 983-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...