N-Boc-3-trifluoromethyl-L-phenylalanine 97%

Supplier: Ambeed
Warning

A746011-1G A746011-250MG A746011-5G
76791-272EA 61 USD
76791-272 76791-274 76791-276
N-Boc-3-trifluoromethyl-L-phenylalanine 97%
N-Boc-3-trifluoromethyl-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₅H₁₈F₃NO₄
MDL Number: MFCD00672524
CAS Number: 142995-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR