4,5-Dicarboxy-γ-pentadecanolactone 98%

Supplier: Ambeed
Warning

A744572-1G A744572-5G
76791-268EA 11 USD
76791-268 76791-270
4,5-Dicarboxy-γ-pentadecanolactone 98%
4,5-Dicarboxy-gamma-pentadecanolactone

Precaución: for Research Use Only

Formula: C₁₇H₂₈O₆
MDL Number: MFCD00059059
CAS Number: 469-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR