3-Isopropoxybenzoic acid 97%

Supplier: Ambeed
Warning

A739799-1G A739799-5G
76791-246EA 42 USD
76791-246 76791-248
3-Isopropoxybenzoic acid 97%
3-Isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
MDL Number: MFCD01922170
CAS Number: 60772-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR