4-Methoxy-2-(trifluoromethyl)benzonitrile 97%

Supplier: Ambeed
Warning

A739419-10G A739419-250MG A739419-25G
76791-240EA 76.39 USD
76791-240 76791-242 76791-244
4-Methoxy-2-(trifluoromethyl)benzonitrile 97%
4-Methoxy-2-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₆F₃NO
MW: 201.15 g/mol
MDL Number: MFCD06797342
CAS Number: 875664-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR