4-Chloro-3-(cyclopropylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-3-(cyclopropylcarbamoyl)benzeneboronic acid, (4-Chloro-3-(cyclopropylcarbamoyl)phenyl)boronic acid

A735791-100MG A735791-1G A735791-250MG
76791-230EA 37.14 USD
76791-230 76791-232 76791-234
4-Chloro-3-(cyclopropylcarbamoyl)phenylboronic acid 98%
4-Chloro-3-(cyclopropylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BClNO₃
MW: 239.46 g/mol
MDL Number: MFCD07783865
CAS Number: 871332-73-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR