Varenicline tartrate 98%

Supplier: Ambeed
Danger

Synonyms: Chantix, 7,8,9,10-Tetrahydro-6H-6,10-methanoazepino[4,5-g]quinoxaline (2R,3R)-2,3-dihydroxysuccinate

A733852-10MG A733852-1MG A733852-25MG A733852-5MG A733852-1G A733852-5G A733852-250MG A733852-100MG
76791-222EA 19 USD
76791-222 76791-224 76791-226 76791-228 77573-360 77602-890 77669-976 77873-156
Varenicline tartrate 98%
Varenicline tartrate

Precaución: for Research Use Only

Formula: C₁₃H₁₃N₃·C₄H₆O₆
MDL Number: MFCD08460603
CAS Number: 375815-87-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR