Tris(acetonitrile)pentamethylcyclopentadienylruthenium(II) hexafluorophosphate 98%

Supplier: Ambeed
Warning

Synonyms: Pentamethylcyclopentadienyltris (acetonitrile)ruthenium(II) hexafluorophosphate

A731651-100MG A731651-250MG A731651-50MG A731651-1G
76791-210EA 127 USD
76791-210 76791-212 76791-214 77669-968
Tris(acetonitrile)pentamethylcyclopentadienylruthenium(II) hexafluorophosphate 98%
Tris(acetonitrile)pentamethylcyclopentadienylruthenium(II) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₃PRuF₆
MW: 504.4200 g/mol
MDL Number: MFCD04117715
CAS Number: 99604-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR