1-(4-(((2R,4S)-2-((1H-1,2,4-Triazol-1-yl)methyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl)methoxy)phenyl)-4-isopropylpiperazine 97%

Supplier: Ambeed
Warning

Synonyms: Terconazole, 1-[4-[[(2R,4S)-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-4-(1-methylethyl)-rel-piperazine

A728990-100MG A728990-10MG A728990-1G A728990-1MG A728990-250MG A728990-50MG
76791-196EA 49.87 USD
76791-196 76791-198 76791-200 76791-202 76791-204 76791-206
1-(4-(((2R,4S)-2-((1H-1,2,4-Triazol-1-yl)methyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl)methoxy)phenyl)-4-isopropylpiperazine 97%
1-(4-(((2R,4S)-2-((1H-1,2,4-Triazol-1-yl)methyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl)methoxy)phenyl)-4-isopropylpiperazine

Precaución: for Research Use Only

Formula: C₂₆H₃₁Cl₂N₅O₃
MW: 532.46 g/mol
MDL Number: MFCD05662369
CAS Number: 67915-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR