(1R,2S)-2-Amino-1-(Boc-amino)cyclopentane 98%

Supplier: Ambeed
Warning

A728420-100MG A728420-1G
76791-192EA 67.9 USD
76791-192 76791-194
(1R,2S)-2-Amino-1-(Boc-amino)cyclopentane 98%
(1R,2S)-2-Amino-1-(Boc-amino)cyclopentane

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.27 g/mol
MDL Number: MFCD12828694
CAS Number: 721395-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR