Tetramethylammonium fluoride tetrahydrate 98%

Supplier: Ambeed
Warning

A727378-100G A727378-25G A727378-500G A727378-5G
76791-172EA 23 USD
76791-172 76791-174 76791-176 76791-178
Tetramethylammonium fluoride tetrahydrate 98%
Tetramethylammonium fluoride tetrahydrate

Precaución: for Research Use Only

Formula: C₄H₁₂FN·4H₂O
MW: 165.21 g/mol
MDL Number: MFCD00149968
CAS Number: 17787-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR