(3-Chloro-4-methoxyphenyl)methanol 97%

Supplier: Ambeed
Warning

A726979-1G A726979-250MG A726979-5G
76791-166EA 34 USD
76791-166 76791-168 76791-170
(3-Chloro-4-methoxyphenyl)methanol 97%
(3-Chloro-4-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
MDL Number: MFCD04124803
CAS Number: 14503-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR