Zinc (II) phthalocyanine (Sulfate ionophore II) ≥97% (dye content)

Supplier: Ambeed
Warning

Synonyms: Zinc phthalocyanine, 29H,31H-Phthalocyanine, zinc complex

A726791-5G A726791-1G A726791-25G
76791-164EA 30 USD
76791-164 77602-876 77669-960
Zinc (II) phthalocyanine (Sulfate ionophore II) ≥97% (dye content)
Zinc (II) phthalocyanine (Sulphate ionophore II)

Precaución: for Research Use Only

Formula: C₃₂H₁₆N₈Zn
MW: 577.91 g/mol
MDL Number: MFCD00041955
CAS Number: 14320-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR