Tetrakis(dimethylamino)diboron 98%

Supplier: Ambeed
Warning

Synonyms: Tetrakis(dimethylamino)diborane

A725016-100G A725016-25G A725016-5G
76791-154EA 175.05 USD
76791-154 76791-156 76791-158
Tetrakis(dimethylamino)diboron 98%
Tetrakis(dimethylamino)diboron

Precaución: for Research Use Only

Formula: C₈H₂₄B₂N₄
MW: 197.93 g/mol
MDL Number: MFCD00048012
CAS Number: 1630-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR