3-Bromo-5-ethylaniline 98%

Supplier: Ambeed
Danger

A724059-100MG A724059-1G A724059-250MG A724059-5G
76791-146EA 20.16 USD
76791-146 76791-148 76791-150 76791-152
3-Bromo-5-ethylaniline 98%
3-Bromo-5-ethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
MDL Number: MFCD16659612
CAS Number: 123158-68-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR