trans-4-(Boc-amino)cyclohexanecarboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl trans-4-formylcyclohexylcarbamate

A717776-10G A717776-1G A717776-5G A717776-100MG
76791-128EA 264 USD
76791-128 76791-130 77669-930 77873-129
trans-4-(Boc-amino)cyclohexanecarboxaldehyde 97%
trans-4-(Boc-amino)cyclohexanecarboxaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MDL Number: MFCD03844598
CAS Number: 181308-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR