5-Fluoro-2-methoxynicotinaldehyde 95%

Supplier: Ambeed
Warning

A715833-100G A715833-10G A715833-1G A715833-25G A715833-5G
76791-110EA 819 USD
76791-110 76791-112 76791-114 76791-116 76791-118
5-Fluoro-2-methoxynicotinaldehyde 95%
5-Fluoro-2-methoxynicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD04972398
CAS Number: 351410-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR