5-Bromo-1-methyl-1H-indole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1H-Indole-3-carboxylic acid, 5-bromo-1-methyl-, 5-bromo-1-methyl-1H-indole-3-carboxylicacid, 5-Bromo-1-methylindole-3-carboxylic acid

A700371-1G A700371-250MG A700371-5G
76791-070EA 74.27 USD
76791-070 76791-072 76791-074
5-Bromo-1-methyl-1H-indole-3-carboxylic acid 97%
5-Bromo-1-methyl-1H-indole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MW: 254.083 g/mol
MDL Number: MFCD14706162
CAS Number: 400071-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR