(2-Bromo-3-methylphenyl)methanol 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3-methylbenzyl Alcohol

A699860-100MG A699860-1G A699860-250MG A699860-5G
76791-062EA 8 USD
76791-062 76791-064 76791-066 76791-068
(2-Bromo-3-methylphenyl)methanol 95%
(2-Bromo-3-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MDL Number: MFCD16659614
CAS Number: 168886-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR