4-Pentyl-4'-iodobiphenyl 95%
Supplier: Ambeed
|
Warning
|
A696444-100MG
A696444-1G
A696444-250MG
A696444-5G
76791-054EA
5
USD
76791-054
76791-056
76791-058
76791-060
4-Pentyl-4'-iodobiphenyl 95%
4-Pentyl-4'-iodobiphenyl
Precaución: for Research Use Only
Formula:
C₁₇H₁₉I |
MDL Number:
MFCD00445195 CAS Number: 69971-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...