Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate 98%

Supplier: Ambeed
Warning

Synonyms: TRIS(ACETONITRILE)CYCLOPENTADIENYLRUTHENIUM(I) HEXACHLOROPHOSPHATE, Tris(acetonitrile)cyclopentadienylruthenium hexafluorophosphate, Trisacetonitrilecyclopentadienylrutheniumhexachlorophosphate

A695955-250MG A695955-1G A695955-5G
76791-042EA 162 USD
76791-042 77569-324 77873-093
Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate 98%
Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate

Precaución: For research use only.

Formula: C₁₁H₉F₆N₃PRu
MW: 429.24 g/mol
MDL Number: MFCD02684569
CAS Number: 80049-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR