(2-Methoxy-5-methylpyridin-3-yl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-5-methylpyridine-3-boronic acid

A692020-1G A692020-250MG A692020-5G
76791-022EA 189 USD
76791-022 76791-024 76791-026
(2-Methoxy-5-methylpyridin-3-yl)boronic acid 98%
(2-Methoxy-5-methylpyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₃
MDL Number: MFCD07437974
CAS Number: 1029654-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR