(3-Bromo-2-fluorophenyl)acetonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 3-Bromo-2-fluorophenylacetonitrile

A689944-100MG A689944-10G A689944-1G A689944-250MG A689944-25G A689944-5G
76791-010EA 12.74 USD
76791-010 76791-012 76791-014 76791-016 76791-018 76791-020
(3-Bromo-2-fluorophenyl)acetonitrile 98%
(3-Bromo-2-fluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.04 g/mol
MDL Number: MFCD09864662
CAS Number: 874285-03-7
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR