Tri(1-naphthyl)phosphine 97%

Supplier: Ambeed
Warning

Synonyms: Tri(naphthalen-1-yl)phosphine

A685466-10G A685466-1G A685466-25G A685466-5G
76790-996EA 135.8 USD
76790-996 76790-998 76791-000 76791-002
Tri(1-naphthyl)phosphine 97%
Tri(1-naphthyl)phosphine

Precaución: for Research Use Only

Formula: C₃₀H₂₁P
MW: 412.46 g/mol
MDL Number: MFCD00046400
CAS Number: 3411-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR