Triphenylsulfonium trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

A683636-1G A683636-250MG A683636-5G
76790-980EA 59 USD
76790-980 76790-982 76790-984
Triphenylsulfonium trifluoromethanesulfonate 98%
Triphenylsulfonium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₉H₁₅F₃O₃S₂
MW: 412.44 g/mol
MDL Number: MFCD01940916
CAS Number: 66003-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR