4-Amino-2-chloro-3-methylbenzonitrile 97%

Supplier: Ambeed
Warning

A680839-100MG A680839-250MG
76790-976EA 67.9 USD
76790-976 76790-978
4-Amino-2-chloro-3-methylbenzonitrile 97%
4-Amino-2-chloro-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MW: 166.60 g/mol
MDL Number: MFCD08059126
CAS Number: 573768-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR